Assay Details
Target Gene Details
Entrez Gene ID: | 81849 |
Gene Name: | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5 |
Gene Aliases: |
SIAT7-E, SIAT7E, ST6GalNAcV |
Location: |
Chr.1:76867408-77069167 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST6GALNAC5 | NM_001320273.1 | NP_001307202.1 | ||
| NM_001320274.1 | NP_001307203.1 | |||
| NM_030965.2 | NP_112227.1 | |||
| AJ507292.1 | CAD45372.1 | |||
| AK056241.1 | BAB71127.1 | |||
| AK095758.1 | ||||
| AK295019.1 | ||||
| AK300902.1 | ||||
| BC001201.1 | AAH01201.1 | |||
| DA166913.1 | ||||
| DC358960.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005649 | Chr.1:76675354 - 76933062 on Build GRCh38 | Loss |
|
| nsv1003386 | Chr.1:76839512 - 76909851 on Build GRCh38 | Gain |
|
| esv3581804 | Chr.1:76831507 - 76916035 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79508697] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map