Assay Details
Target Gene Details
Entrez Gene ID: | 5629 |
Gene Name: | prospero homeobox 1 |
Gene Aliases: |
- |
Location: |
Chr.1:213983239-214041510 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROX1 | XM_011509773.2 | XP_011508075.1 |
Target Gene Details
Entrez Gene ID: | 100505832 |
Gene Name: | PROX1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:213819641-213986153 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROX1-AS1 | NR_037850.2 | |||
| AV681804.1 | ||||
| BE179719.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952088 | Chr.1:213980058 - 213991157 on Build GRCh38 | Deletion |
|
| nsv4498 | Chr.1:213981463 - 214016219 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78915050] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map