Assay Details
Target Gene Details
Entrez Gene ID: | 388646 |
Gene Name: | guanylate binding protein 7 |
Gene Aliases: |
GBP4L |
Location: |
Chr.1:89131751-89176040 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBP7 | NM_207398.2 | NP_997281.2 | ||
| AK096141.1 | BAC04709.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830548 | Chr.1:89142066 - 89298597 on Build GRCh38 | Gain |
|
| esv2714562 | Chr.1:89133415 - 89186630 on Build GRCh38 | Deletion |
|
| nsv546796 | Chr.1:89052437 - 89256179 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map