Assay Details
Target Gene Details
Entrez Gene ID: | 9132 |
Gene Name: | potassium voltage-gated channel subfamily Q member 4 |
Gene Aliases: |
DFNA2, DFNA2A, KV7.4 |
Location: |
Chr.1:40784012-40840457 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ4 | NM_004700.3 | NP_004691.2 | ||
| NM_172163.2 | NP_751895.1 | |||
| AF105202.1 | AAD14680.1 |
More Information
Set Membership: |
|

Genomic Map