Assay Details
Target Gene Details
Entrez Gene ID: | 101928068 |
Gene Name: | uncharacterized LOC101928068 |
Gene Aliases: |
- |
Location: |
Chr.1:244965204-244970869 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928068 | NR_111907.1 | 2 | 1181 | |
| AK126649.1 | 1 | 198 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589420 | Chr.1:244890996 - 245040215 on Build GRCh38 | Loss |
|
| nsv5188 | Chr.1:244962548 - 244994285 on Build GRCh38 | Insertion |
|
| nsv471922 | Chr.1:244968832 - 244968832 on Build GRCh38 | Novel Sequence Insertion |
|
| esv22057 | Chr.1:244894679 - 245033598 on Build GRCh38 | Loss |
|
| dgv602n100 | Chr.1:244936197 - 245539982 on Build GRCh38 | Gain |
|

Genomic Map