Assay Details
Target Gene Details
Entrez Gene ID: | 29956 |
Gene Name: | ceramide synthase 2 |
Gene Aliases: |
L3, LASS2, SP260, TMSG1 |
Location: |
Chr.1:150965173-150975003 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERS2 | NM_022075.4 | NP_071358.1 | ||
| NM_181746.3 | NP_859530.1 | |||
| XM_011509451.2 | XP_011507753.1 | |||
| XM_011509452.2 | XP_011507754.1 | |||
| AF177338.2 | AAG17982.2 | |||
| AK001105.1 | BAA91505.1 | |||
| AK290989.1 | ||||
| AY091458.1 | AAM12028.1 | |||
| BC001357.2 | AAH01357.2 | |||
| BC010032.2 | AAH10032.1 | |||
| BM764143.1 | ||||
| BQ219926.1 | ||||
| BU543940.1 | ||||
| DA019883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522317 | Chr.1:150961247 - 151001686 on Build GRCh38 | Loss |
|
| nsv952804 | Chr.1:150964125 - 150976324 on Build GRCh38 | Deletion |
|
| nsv1008496 | Chr.1:150873351 - 150998180 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map