Assay Details
Target Gene Details
Entrez Gene ID: | 79947 |
Gene Name: | dehydrodolichyl diphosphate synthase subunit |
Gene Aliases: |
CIT, CPT, DS, HDS, RP59 |
Location: |
Chr.1:26432282-26471306 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Target Gene Details
Entrez Gene ID: | 101928324 |
Gene Name: | uncharacterized LOC101928324 |
Gene Aliases: |
- |
Location: |
Chr.1:26462756-26467537 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928324 | NR_125952.1 | |||
| NR_125953.1 | ||||
| AL050085.1 | ||||
| BF511893.1 |
More Information
Set Membership: |
|

Genomic Map