Assay Details
Target Gene Details
Entrez Gene ID: | 55225 |
Gene Name: | ribonucleoprotein, PTB binding 2 |
Gene Aliases: |
- |
Location: |
Chr.1:64744979-64833232 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAVER2 | NM_018211.3 | NP_060681.2 | ||
| XM_006710738.3 | XP_006710801.2 | |||
| XM_011541706.2 | XP_011540008.1 | |||
| AB046799.1 |
More Information
Set Membership: |
|

Genomic Map