Assay Details
Target Gene Details
Entrez Gene ID: | 126969 |
Gene Name: | solute carrier family 44 member 3 |
Gene Aliases: |
CTL3 |
Location: |
Chr.1:94820342-94895247 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
More Information
Set Membership: |
|

Genomic Map