Assay Details
Target Gene Details
Entrez Gene ID: | 127294 |
Gene Name: | myomesin 3 |
Gene Aliases: |
- |
Location: |
Chr.1:24056041-24112175 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOM3 | NM_152372.3 | NP_689585.3 | ||
| AK128233.1 | BAC87343.1 | |||
| BC067101.1 | AAH67101.1 | |||
| BX537911.1 | CAD97895.1 | |||
| BX647453.1 | CAH10377.1 | |||
| BX647455.1 | CAH10376.1 | |||
| BX647644.1 |
More Information
Set Membership: |
|

Genomic Map