Assay Details
Target Gene Details
Entrez Gene ID: | 54996 |
Gene Name: | mitochondrial amidoxime reducing component 2 |
Gene Aliases: |
MOSC2 |
Location: |
Chr.1:220748246-220784815 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MARC2 | NM_001317338.1 | NP_001304267.1 | ||
| NM_017898.4 | NP_060368.2 | |||
| XM_006711407.2 | XP_006711470.1 | |||
| XM_011509684.1 | XP_011507986.1 | |||
| XM_017001581.1 | XP_016857070.1 | |||
| AK000612.1 | BAA91287.1 | |||
| AL136931.1 | CAB66865.1 | |||
| BC011973.1 | AAH11973.1 | |||
| BC015829.2 | AAH15829.2 | |||
| BC016859.1 | AAH16859.1 | |||
| BP194343.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523389 | Chr.1:220753502 - 220782670 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72472389] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map