Assay Details
Target Gene Details
Entrez Gene ID: | 54558 |
Gene Name: | spermatogenesis associated 6 |
Gene Aliases: |
HASH, SRF-1, SRF1 |
Location: |
Chr.1:48258643-48472208 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATA6 | XM_006710699.2 | 13 | 4347 | XP_006710762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011588 | Chr.1:48248029 - 48328196 on Build GRCh38 | Gain |
|
| esv3578249 | Chr.1:48202315 - 48332128 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map