Assay Details
Target Gene Details
Entrez Gene ID: | 3814 |
Gene Name: | KiSS-1 metastasis-suppressor |
Gene Aliases: |
HH13, KiSS-1 |
Location: |
Chr.1:204190341-204196491 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KISS1 | NM_002256.3 | NP_002247.3 | ||
| AF218017.1 | AAG17259.1 | |||
| AK291695.1 | ||||
| AY117143.1 | AAM78592.1 | |||
| BC022819.1 | AAH22819.1 | |||
| U43527.1 | AAC79512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762138 | Chr.1:204138959 - 204248018 on Build GRCh38 | Gain |
|
| nsv1007228 | Chr.1:204174999 - 204216444 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map