Assay Details
Target Gene Details
Entrez Gene ID: | 140576 |
Gene Name: | S100 calcium binding protein A16 |
Gene Aliases: |
AAG13, DT1P1A7, S100F |
Location: |
Chr.1:153606883-153613168 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S100A16 | NM_080388.2 | NP_525127.1 | ||
NR_133645.1 | ||||
AW954845.1 | ||||
AY221961.1 | AAP46152.1 | |||
AY762098.1 | ||||
BC010541.1 | AAH10541.1 | |||
BC019099.1 | AAH19099.1 | |||
BM739127.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
