Assay Details
Target Gene Details
Entrez Gene ID: | 11123 |
Gene Name: | RCAN family member 3 |
Gene Aliases: |
DSCR1L2, MCIP3, RCN3, hRCN3 |
Location: |
Chr.1:24502351-24537020 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCAN3 | NM_001251977.1 | NP_001238906.1 | ||
| NM_001251979.1 | NP_001238908.1 | |||
| NM_001251981.1 | NP_001238910.1 | |||
| NM_001251983.1 | NP_001238912.1 | |||
| NM_001251984.1 | NP_001238913.1 | |||
| NM_013441.3 | NP_038469.1 | |||
| BC035854.1 | AAH35854.1 | |||
| BP326714.1 | ||||
| CT001954.1 | ||||
| CX752875.1 | ||||
| GQ411200.1 | ||||
| HQ287726.1 | ||||
| HQ317421.1 | ||||
| HQ317422.1 | ||||
| JN228252.1 |
More Information
Set Membership: |
|

Genomic Map