Assay Details
Target Gene Details
Entrez Gene ID: | 6232 |
Gene Name: | ribosomal protein S27 |
Gene Aliases: |
MPS-1, MPS1, S27 |
Location: |
Chr.1:153990763-153992155 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS27 | AK312070.1 | |||
| CD177656.1 | ||||
| L19739.1 | AAA59867.1 | |||
| U57847.1 | AAB02266.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv946401 | Chr.1:153991361 - 153992953 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map