Assay Details
Target Gene Details
Entrez Gene ID: | 51102 |
Gene Name: | mitochondrial trans-2-enoyl-CoA reductase |
Gene Aliases: |
CGI-63, FASN2B, NRBF1 |
Location: |
Chr.1:29192657-29230958 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MECR | NM_001024732.2 | NP_001019903.2 | ||
| NM_016011.3 | NP_057095.3 | |||
| H17678.1 | 1 | 305 |
More Information
Set Membership: |
|

Genomic Map