Assay Details
Target Gene Details
Entrez Gene ID: | 100506161 |
Gene Name: | TAF1A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:222589920-222592633 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF1A-AS1 | NR_110613.1 | |||
| NR_110614.1 | ||||
| DA202557.1 | ||||
| HY020214.1 |

Genomic Map