Assay Details
Target Gene Details
Entrez Gene ID: | 6536 |
Gene Name: | solute carrier family 6 member 9 |
Gene Aliases: |
GLYT1 |
Location: |
Chr.1:43996483-44031492 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A9 | NM_001024845.2 | NP_001020016.1 | ||
| NM_001261380.1 | NP_001248309.1 | |||
| NM_006934.3 | NP_008865.2 | |||
| NM_201649.3 | NP_964012.2 | |||
| NR_048548.1 | ||||
| XM_011542017.2 | XP_011540319.1 | |||
| XM_017002151.1 | XP_016857640.1 | |||
| XM_017002152.1 | XP_016857641.1 | |||
| XM_017002153.1 | XP_016857642.1 | |||
| AI884725.1 | ||||
| AK296438.1 | ||||
| AK298611.1 | ||||
| AK304057.1 | ||||
| AK311136.1 | ||||
| AK315233.1 | ||||
| AK315933.1 | ||||
| S70609.1 | AAB30784.1 | |||
| S70612.1 | AAB30785.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75404559] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map