Assay Details
Target Gene Details
Entrez Gene ID: | 732 |
Gene Name: | complement component 8, beta polypeptide |
Gene Aliases: |
C82 |
Location: |
Chr.1:56929210-56974383 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8B | NM_000066.3 | NP_000057.2 | ||
| NM_001278543.1 | NP_001265472.1 | |||
| NM_001278544.1 | NP_001265473.1 | |||
| XM_017002235.1 | XP_016857724.1 | |||
| AK292740.1 | ||||
| AK298425.1 | ||||
| AK298444.1 | ||||
| AK313382.1 | ||||
| AK316209.1 | ||||
| BC020701.1 | ||||
| BC130575.1 | ||||
| M16973.1 | AAA51862.1 | |||
| X04393.1 | CAA27981.1 |
More Information
Set Membership: |
|

Genomic Map