Assay Details
Target Gene Details
Entrez Gene ID: | 6944 |
Gene Name: | vacuolar protein sorting 72 homolog |
Gene Aliases: |
CFL1, Swc2, TCFL1, YL-1, YL1 |
Location: |
Chr.1:151176300-151190213 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS72 | NM_001271087.1 | NP_001258016.1 | ||
| NM_001271088.1 | NP_001258017.1 | |||
| NM_005997.2 | NP_005988.1 | |||
| XM_017002205.1 | XP_016857694.1 | |||
| AK222935.1 | BAD96655.1 | |||
| AK222939.1 | BAD96659.1 | |||
| AK225130.1 | ||||
| AK297565.1 | ||||
| AK310045.1 | ||||
| BC003151.1 | AAH03151.1 | |||
| BT019356.1 | AAV38163.1 | |||
| BT019357.1 | AAV38164.1 | |||
| CT003766.1 | ||||
| CX163016.1 | ||||
| D43642.1 | BAA07757.1 | |||
| DQ655941.2 | ||||
| DQ656047.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map