Assay Details
Target Gene Details
Entrez Gene ID: | 22955 |
Gene Name: | sex comb on midleg homolog 1 (Drosophila) |
Gene Aliases: |
Scml3 |
Location: |
Chr.1:41027199-41242155 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546103 | Chr.1:41011772 - 41076832 on Build GRCh38 | Loss |
|
| esv3585819 | Chr.1:41049509 - 41053926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map