Assay Details
Target Gene Details
Entrez Gene ID: | 149420 |
Gene Name: | PDLIM1 interacting kinase 1 like |
Gene Aliases: |
CLIK1L, STK35L2 |
Location: |
Chr.1:26109406-26125548 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDIK1L | XM_006710381.3 | 1 | 1275 | XP_006710444.1 |
More Information
Set Membership: |
|

Genomic Map