Assay Details
Target Gene Details
Entrez Gene ID: | 51043 |
Gene Name: | zinc finger and BTB domain containing 7B |
Gene Aliases: |
CKROX, THPOK, ZBTB15, ZFP-67, ZFP67, ZNF857B, c-KROX, hcKROX |
Location: |
Chr.1:155002628-155018525 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB7B | NM_001252406.2 | NP_001239335.1 | ||
| NM_001256455.1 | NP_001243384.1 | |||
| NR_045515.1 | ||||
| NR_046206.1 | ||||
| NR_049765.1 | ||||
| XM_006711349.2 | XP_006711412.2 | |||
| XM_006711353.1 | XP_006711416.1 | |||
| XM_006711354.1 | XP_006711417.1 | |||
| XM_006711356.3 | XP_006711419.1 | |||
| XM_006711357.1 | XP_006711420.1 | |||
| XM_006711358.1 | XP_006711421.1 | |||
| XM_006711359.2 | XP_006711422.1 | |||
| XM_011509598.2 | XP_011507900.1 | |||
| XM_011509599.2 | XP_011507901.1 | |||
| XM_017001399.1 | XP_016856888.1 | |||
| AK291619.1 | ||||
| AK304762.1 | ||||
| BC012070.1 | AAH12070.1 | |||
| BF510901.1 | ||||
| BM801567.1 | ||||
| CR749303.1 | CAH18158.1 | |||
| CV572166.1 | ||||
| DA970906.1 | ||||
| DB241444.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
| nsv952212 | Chr.1:154998225 - 155087624 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79174338,rs80334315] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map