Assay Details
Target Gene Details
Entrez Gene ID: | 3756 |
Gene Name: | potassium voltage-gated channel subfamily H member 1 |
Gene Aliases: |
EAG, EAG1, Kv10.1, TMBTS, ZLS1, h-eag, hEAG1 |
Location: |
Chr.1:210678314-211134115 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNH1 | NM_002238.3 | 11 | 5084 | NP_002229.1 |
| NM_172362.2 | 11 | 5165 | NP_758872.1 | |
| XM_017001246.1 | 6 | 4303 | XP_016856735.1 | |
| AK127526.1 | 1 | 677 |
More Information
Set Membership: |
|

Genomic Map