Assay Details
Target Gene Details
Entrez Gene ID: | 58155 |
Gene Name: | polypyrimidine tract binding protein 2 |
Gene Aliases: |
PTBLP, brPTB, nPTB |
Location: |
Chr.1:96721605-96815049 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTBP2 | NM_001300985.1 | NP_001287914.1 | ||
| NM_001300986.1 | NP_001287915.1 | |||
| NM_001300987.1 | NP_001287916.1 | |||
| NM_001300988.1 | NP_001287917.1 | |||
| NM_001300989.1 | NP_001287918.1 | |||
| NM_001300990.1 | NP_001287919.1 | |||
| NM_021190.3 | NP_067013.1 | |||
| NR_125356.1 | ||||
| NR_125357.1 | ||||
| XM_011541874.2 | XP_011540176.1 | |||
| XM_011541875.2 | XP_011540177.1 | |||
| XM_011541876.2 | XP_011540178.1 | |||
| XM_017001951.1 | XP_016857440.1 | |||
| AB051232.1 | BAB71742.1 | |||
| AB051233.1 | BAB71743.1 | |||
| AB209266.1 | BAD92503.1 | |||
| AF176085.1 | AAF14284.1 | |||
| AF530580.1 | AAM94624.1 | |||
| AF530581.1 | AAM94625.1 | |||
| AK092216.1 | ||||
| AK295384.1 | ||||
| AK299755.1 | ||||
| AK299897.1 | ||||
| AK299922.1 | ||||
| BC016582.2 | AAH16582.1 | |||
| BP250235.1 | ||||
| DB473562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830681 | Chr.1:96700230 - 96872508 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map