Assay Details
Target Gene Details
Entrez Gene ID: | 57554 |
Gene Name: | leucine rich repeat containing 7 |
Gene Aliases: |
DENSIN |
Location: |
Chr.1:69567130-70152426 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC7 | XM_017001890.1 | 1 | 1385 | XP_016857379.1 |
| XM_017001891.1 | 1 | 1385 | XP_016857380.1 | |
| AF498274.1 | 1 | 65 | AAP06801.1 | |
| AK294206.1 | 1 | 242 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830126 | Chr.1:69549196 - 69702296 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map