Assay Details
Target Gene Details
Entrez Gene ID: | 51246 |
Gene Name: | shisa family member 5 |
Gene Aliases: |
SCOTIN |
Location: |
Chr.3:48467798-48504826 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHISA5 | NM_001272065.1 | NP_001258994.1 | ||
| NM_001272066.1 | NP_001258995.1 | |||
| NM_001272067.1 | NP_001258996.1 | |||
| NM_001272068.1 | NP_001258997.1 | |||
| NM_016479.4 | NP_057563.3 | |||
| XM_005265205.1 | XP_005265262.1 | |||
| XM_006713188.2 | XP_006713251.1 | |||
| XM_011533796.1 | XP_011532098.1 | |||
| AF520698.1 | AAM74232.1 | |||
| AK056328.1 | BAB71152.1 | |||
| AK075441.1 | BAC11623.1 | |||
| AK124652.1 | ||||
| AK301191.1 | ||||
| BI552637.1 | ||||
| BX336842.2 | ||||
| BX344125.2 | ||||
| BX379507.2 | ||||
| DA025113.1 | ||||
| DA045437.1 | ||||
| DA657994.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834684 | Chr.3:48344209 - 48551890 on Build GRCh38 | Gain |
|
| nsv3809 | Chr.3:48474359 - 48502729 on Build GRCh38 | Insertion |
|
| dgv8315n54 | Chr.3:48464432 - 48478412 on Build GRCh38 | Loss |
|
| nsv590242 | Chr.3:48392157 - 48518906 on Build GRCh38 | Loss |
|
| nsv508917 | Chr.3:48400224 - 48499357 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map