Assay Details
Target Gene Details
Entrez Gene ID: | 4241 |
Gene Name: | melanotransferrin |
Gene Aliases: |
CD228, MAP97, MFI2, MTF1, MTf |
Location: |
Chr.3:197001740-197029816 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MELTF | NM_005929.5 | NP_005920.2 | ||
| NM_033316.3 | NP_201573.1 | |||
| XM_006713643.3 | XP_006713706.2 | |||
| XM_011512850.2 | XP_011511152.1 | |||
| XM_011512851.2 | XP_011511153.1 | |||
| A00127.1 | CAA00012.1 | |||
| AA442552.1 | ||||
| AU099352.1 | ||||
| BC001875.1 | AAH01875.1 | |||
| BC002623.2 | AAH02623.1 | |||
| BC007550.1 | AAH07550.1 | |||
| BC071910.1 | AAH71910.1 | |||
| BC093920.1 | AAH93920.1 | |||
| BC111947.1 | AAI11948.1 | |||
| BT007377.1 | AAP36041.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599293 | Chr.3:196759100 - 197444213 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map