Assay Details
Target Gene Details
Entrez Gene ID: | 285352 |
Gene Name: | KIF9 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:47164370-47244116 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF9-AS1 | NR_033373.1 | |||
| AK096853.1 | ||||
| DA755837.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1007714 | Chr.3:47042897 - 47290080 on Build GRCh38 | Gain |
|
| nsv3801 | Chr.3:47160040 - 47204948 on Build GRCh38 | Deletion |
|
| nsv590211 | Chr.3:47147995 - 47214567 on Build GRCh38 | Gain |
|
| dgv1373e212 | Chr.3:47191381 - 47209431 on Build GRCh38 | Gain |
|
| nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
|
| esv3596077 | Chr.3:47192046 - 47215375 on Build GRCh38 | Gain |
|
| esv3391060 | Chr.3:47200158 - 47203156 on Build GRCh38 | Duplication |
|

Genomic Map