Assay Details
Target Gene Details
Entrez Gene ID: | 23132 |
Gene Name: | RAD54-like 2 (S. cerevisiae) |
Gene Aliases: |
ARIP4, HSPC325, SRISNF2L |
Location: |
Chr.3:51538683-51668660 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD54L2 | NM_001322253.1 | NP_001309182.1 | ||
| NM_001322256.1 | NP_001309185.1 | |||
| NM_015106.3 | NP_055921.2 | |||
| XM_017005988.1 | 1 | 1008 | XP_016861477.1 | |
| HY047442.1 | ||||
| HY049223.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999553 | Chr.3:51522568 - 51662036 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map