Assay Details
Target Gene Details
Entrez Gene ID: | 22908 |
Gene Name: | SAC1 suppressor of actin mutations 1-like (yeast) |
Gene Aliases: |
SAC1 |
Location: |
Chr.3:45689241-45745425 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SACM1L | NM_001319072.1 | NP_001306001.1 | ||
| NM_001319073.1 | NP_001306002.1 | |||
| NM_014016.4 | NP_054735.3 | |||
| XM_011533500.2 | XP_011531802.1 | |||
| AB020658.1 | ||||
| AK291142.1 | ||||
| AK296421.1 | ||||
| AK301248.1 | ||||
| AL136831.1 | CAB66765.1 | |||
| BC016559.1 | AAH16559.1 | |||
| CR749564.1 | CAH18361.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34130 | Chr.3:45567583 - 45755165 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map