Assay Details
Target Gene Details
Entrez Gene ID: | 285331 |
Gene Name: | coiled-coil domain containing 66 |
Gene Aliases: |
- |
Location: |
Chr.3:56557125-56621837 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460557 | Chr.3:56560707 - 56627292 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map