Assay Details
Target Gene Details
Entrez Gene ID: | 9922 |
Gene Name: | IQ motif and Sec7 domain 1 |
Gene Aliases: |
ARF-GEP100, ARFGEP100, BRAG2, GEP100 |
Location: |
Chr.3:12897043-13282998 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQSEC1 | NM_001134382.2 | NP_001127854.1 | ||
| NM_014869.6 | NP_055684.3 | |||
| XM_011534304.2 | XP_011532606.1 | |||
| XM_011534305.2 | XP_011532607.1 | |||
| XM_011534306.2 | XP_011532608.1 | |||
| XM_011534307.2 | XP_011532609.1 | |||
| XM_011534308.2 | XP_011532610.1 | |||
| XM_011534310.2 | XP_011532612.1 | |||
| XM_011534311.2 | XP_011532613.1 | |||
| XM_011534312.2 | XP_011532614.1 | |||
| XM_011534313.2 | XP_011532615.1 | |||
| XM_011534314.2 | XP_011532616.1 | |||
| XM_011534315.2 | XP_011532617.1 | |||
| XM_017007581.1 | XP_016863070.1 | |||
| XM_017007582.1 | XP_016863071.1 | |||
| AB018306.1 | BAA34483.2 | |||
| AK294525.1 | ||||
| AY653734.1 | AAT72063.1 | |||
| BC010267.1 | AAH10267.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589716 | Chr.3:12820109 - 12926604 on Build GRCh38 | Loss |
|
| nsv589715 | Chr.3:12813127 - 12976988 on Build GRCh38 | Loss |
|
| nsv589718 | Chr.3:12898047 - 12960142 on Build GRCh38 | Loss |
|
| esv3893663 | Chr.3:12822887 - 12921716 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map