Assay Details
Target Gene Details
Entrez Gene ID: | 170572 |
Gene Name: | 5-hydroxytryptamine receptor 3C |
Gene Aliases: |
- |
Location: |
Chr.3:184053047-184060673 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR3C | NM_130770.2 | NP_570126.2 | ||
| AF459285.1 | AAL66182.1 | |||
| BC131799.1 |
More Information
Set Membership: |
|

Genomic Map