Assay Details
Target Gene Details
Entrez Gene ID: | 7348 |
Gene Name: | uroplakin 1B |
Gene Aliases: |
TSPAN20, UPIB, UPK1 |
Location: |
Chr.3:119173578-119205153 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UPK1B | NM_006952.3 | NP_008883.2 | ||
| AB002155.1 | BAA88878.1 | |||
| AK303924.1 | ||||
| AK313787.1 | ||||
| AL709739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597552 | Chr.3:119137605 - 119207994 on Build GRCh38 | Loss |
|
| esv25249 | Chr.3:119136490 - 119281404 on Build GRCh38 | Loss |
|
| esv3597553 | Chr.3:119144924 - 119291446 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map