Assay Details
Target Gene Details
Entrez Gene ID: | 132204 |
Gene Name: | synaptoporin |
Gene Aliases: |
SPO |
Location: |
Chr.3:63200604-63616924 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNPR | NM_001130003.1 | NP_001123475.1 | ||
| NM_144642.4 | NP_653243.1 | |||
| XM_017005731.1 | XP_016861220.1 | |||
| XM_017005732.1 | XP_016861221.1 | |||
| AF411860.2 | AAN03681.2 | |||
| AK122831.1 | ||||
| AK293628.1 | ||||
| AK309017.1 | ||||
| AK312465.1 | ||||
| AK315958.1 | ||||
| AL834457.1 | CAD39117.1 | |||
| BC022518.1 | AAH22518.1 | |||
| CR936762.1 | ||||
| DA505318.1 |
Target Gene Details
Entrez Gene ID: | 100874016 |
Gene Name: | SYNPR antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:63423596-63550051 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNPR-AS1 | NR_046677.1 | |||
| BU959256.1 |
More Information
Set Membership: |
|

Genomic Map