Assay Details
Target Gene Details
Entrez Gene ID: | 2803 |
Gene Name: | golgin A4 |
Gene Aliases: |
CRPF46, GCP2, GOLG, MU-RMS-40.18, p230 |
Location: |
Chr.3:37243191-37366879 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834665 | Chr.3:37180973 - 37368088 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map