Assay Details
Target Gene Details
Entrez Gene ID: | 101928085 |
Gene Name: | ERICH6 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:150703564-150720146 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERICH6-AS1 | NR_121674.1 | |||
| NR_121675.1 | ||||
| BP431953.1 | ||||
| DA374305.1 | ||||
| DB324976.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs367854897] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map