Assay Details
Target Gene Details
Entrez Gene ID: | 64422 |
Gene Name: | autophagy related 3 |
Gene Aliases: |
APG3, APG3-LIKE, APG3L, PC3-96 |
Location: |
Chr.3:112532507-112561963 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATG3 | NM_001278712.1 | NP_001265641.1 | ||
| NM_022488.4 | NP_071933.2 | |||
| XM_011513074.1 | XP_011511376.1 | |||
| AA448974.1 | ||||
| AB079384.1 | BAB90843.1 | |||
| AF202092.1 | AAG35611.1 | |||
| AK307432.1 | ||||
| AK308919.1 | ||||
| AL137515.1 | CAB70781.1 | |||
| AW408464.1 | ||||
| BC002830.1 | AAH02830.1 | |||
| BC024221.2 | AAH24221.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591293 | Chr.3:112266260 - 113293950 on Build GRCh38 | Loss |
|
| dgv4840n100 | Chr.3:112407084 - 112559957 on Build GRCh38 | Gain |
|
| nsv1005323 | Chr.3:112453143 - 112555766 on Build GRCh38 | Gain |
|
| nsv591301 | Chr.3:112533685 - 112613782 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78849600] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map