Assay Details
Target Gene Details
Entrez Gene ID: | 285386 |
Gene Name: | tumor protein p63 regulated 1 |
Gene Aliases: |
FAM79B |
Location: |
Chr.3:189000774-189325304 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPRG1 | XM_017006257.1 | XP_016861746.1 | ||
| XM_017006258.1 | XP_016861747.1 | |||
| XM_017006259.1 | XP_016861748.1 | |||
| XM_017006260.1 | XP_016861749.1 | |||
| XM_017006261.1 | XP_016861750.1 | |||
| AK123232.1 | BAC85565.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599047 | Chr.3:189123548 - 189127296 on Build GRCh38 | Loss |
|

Genomic Map