Assay Details
Target Gene Details
Entrez Gene ID: | 6769 |
Gene Name: | SH3 and cysteine rich domain |
Gene Aliases: |
STAC1 |
Location: |
Chr.3:36380455-36548007 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAC | NM_001292049.1 | NP_001278978.1 | ||
| NM_003149.2 | NP_003140.1 | |||
| XM_011534036.2 | XP_011532338.1 | |||
| XM_011534037.2 | XP_011532339.1 | |||
| XM_011534038.2 | XP_011532340.1 | |||
| AK299160.1 | ||||
| AK302697.1 | ||||
| AK310425.1 | ||||
| AK313493.1 | ||||
| BC020221.1 | AAH20221.1 | |||
| D86640.1 | BAA13152.1 | |||
| DC351652.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3595854 | Chr.3:36329890 - 36407461 on Build GRCh38 | Gain |
|
| esv3595861 | Chr.3:36375559 - 36404508 on Build GRCh38 | Gain |
|
| nsv1006443 | Chr.3:36361829 - 36476267 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map