Assay Details
Target Gene Details
Entrez Gene ID: | 8833 |
Gene Name: | guanine monophosphate synthase |
Gene Aliases: |
- |
Location: |
Chr.3:155869430-155943679 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMPS | XM_011513263.2 | XP_011511565.1 |
More Information
Set Membership: |
|

Genomic Map