Assay Details
Target Gene Details
Entrez Gene ID: | 116933 |
Gene Name: | CLRN1 antisense RNA 1 |
Gene Aliases: |
CLRN1OS, UCRP |
Location: |
Chr.3:150972678-151079830 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLRN1-AS1 | NR_024066.1 | |||
| AF388367.1 | ||||
| AK126442.1 |

Genomic Map