Assay Details
Target Gene Details
Entrez Gene ID: | 7220 |
Gene Name: | transient receptor potential cation channel subfamily C member 1 |
Gene Aliases: |
HTRP-1, TRP1 |
Location: |
Chr.3:142724032-142807888 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC1 | NM_001251845.1 | NP_001238774.1 | ||
| NM_003304.4 | NP_003295.1 | |||
| XM_005247738.3 | XP_005247795.1 | |||
| XM_005247739.2 | XP_005247796.1 | |||
| XM_017007121.1 | XP_016862610.1 | |||
| XM_017007122.1 | XP_016862611.1 | |||
| AB255425.1 | ||||
| BC112338.1 | ||||
| BC113953.1 | ||||
| X89066.1 | CAA61447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv300n21 | Chr.3:142692358 - 142784763 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs146236982] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map