Assay Details
Target Gene Details
Entrez Gene ID: | 590 |
Gene Name: | butyrylcholinesterase |
Gene Aliases: |
CHE1, CHE2, E1 |
Location: |
Chr.3:165772904-165837467 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCHE | XM_017006967.1 | XP_016862456.1 | ||
| AK292063.1 | ||||
| BC008396.1 | AAH08396.1 | |||
| BC018141.1 | AAH18141.1 | |||
| CD014089.1 | ||||
| M16474.1 | AAA52015.1 | |||
| M16541.1 | AAA98113.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8724n54 | Chr.3:165721745 - 165840684 on Build GRCh38 | Loss |
|
| nsv592472 | Chr.3:165464658 - 166106549 on Build GRCh38 | Gain |
|
| esv3598511 | Chr.3:165730641 - 165812290 on Build GRCh38 | Loss |
|
| nsv520596 | Chr.3:165787389 - 166149690 on Build GRCh38 | Gain+Loss |
|
| esv3598513 | Chr.3:165786388 - 165791442 on Build GRCh38 | Loss |
|
| nsv428426 | Chr.3:164382966 - 165879735 on Build GRCh38 | Loss |
|
| dgv4977n100 | Chr.3:165787737 - 165884781 on Build GRCh38 | Loss |
|
| esv3598510 | Chr.3:165729982 - 165943642 on Build GRCh38 | Loss |
|
| esv3893808 | Chr.3:165596587 - 166056006 on Build GRCh38 | Loss |
|
| esv2657931 | Chr.3:165729972 - 165943688 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map