Assay Details
Target Gene Details
Entrez Gene ID: | 151651 |
Gene Name: | EF-hand domain family member B |
Gene Aliases: |
CFAP21 |
Location: |
Chr.3:19879472-19947036 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFHB | NM_144715.3 | NP_653316.3 | ||
| XM_005264889.1 | XP_005264946.1 | |||
| XM_011533382.1 | XP_011531684.1 | |||
| XM_011533383.2 | XP_011531685.2 | |||
| XM_011533385.2 | XP_011531687.1 | |||
| XM_017005741.1 | XP_016861230.1 | |||
| XM_017005742.1 | XP_016861231.1 | |||
| XM_017005743.1 | XP_016861232.1 | |||
| XM_017005744.1 | XP_016861233.1 | |||
| XM_017005745.1 | XP_016861234.1 | |||
| AK097644.1 | ||||
| AK122616.1 | BAC85491.1 | |||
| BC028198.1 | AAH28198.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893667 | Chr.3:19470582 - 20214665 on Build GRCh38 | Gain |
|
| nsv1010123 | Chr.3:18948815 - 19948012 on Build GRCh38 | Gain |
|
| esv35078 | Chr.3:19513779 - 20728244 on Build GRCh38 | Gain |
|
| nsv433358 | Chr.3:19505692 - 19955016 on Build GRCh38 | Gain |
|
| esv23068 | Chr.3:19494161 - 20597009 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73188474] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map