Assay Details
Target Gene Details
Entrez Gene ID: | 57092 |
Gene Name: | PEST proteolytic signal containing nuclear protein |
Gene Aliases: |
- |
Location: |
Chr.3:101574094-101594465 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508944 | Chr.3:101528526 - 101575525 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map