Assay Details
Target Gene Details
Entrez Gene ID: | 7881 |
Gene Name: | potassium voltage-gated channel subfamily A member regulatory beta subunit 1 |
Gene Aliases: |
AKR6A3, KCNA1B, KV-BETA-1, Kvb1.3, hKvBeta3, hKvb3 |
Location: |
Chr.3:156118211-156540603 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNAB1 | NM_001308217.1 | NP_001295146.1 | ||
| NM_001308222.1 | NP_001295151.1 | |||
| NM_003471.3 | NP_003462.2 | |||
| NM_172159.3 | NP_751891.1 | |||
| NM_172160.2 | NP_751892.1 | |||
| XM_011513115.2 | XP_011511417.1 | |||
| XM_017007171.1 | XP_016862660.1 | |||
| XM_017007172.1 | XP_016862661.1 | |||
| XM_017007174.1 | XP_016862663.1 | |||
| AK057059.1 | ||||
| AK127240.1 | ||||
| AK292693.1 | ||||
| AK292999.1 | ||||
| AK303287.1 | ||||
| AY780786.1 | AAV98356.1 | |||
| BC043166.2 | AAH43166.1 | |||
| DA408045.1 | ||||
| DC410397.1 | ||||
| L39833.1 | AAC37573.1 | |||
| L47665.1 | AAC41926.1 | |||
| U16953.1 | AAC50122.1 | |||
| U17968.1 | AAC50113.1 | |||
| U33428.1 | AAC50953.1 |
Target Gene Details
Entrez Gene ID: | 100874084 |
Gene Name: | KCNAB1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:156441157-156446936 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNAB1-AS1 | NR_046618.2 | |||
| BX104028.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76811843] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map