Assay Details
Target Gene Details
Entrez Gene ID: | 152100 |
Gene Name: | C-X9-C motif containing 1 |
Gene Aliases: |
C3orf68 |
Location: |
Chr.3:28241588-28322874 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMC1 | NM_182523.1 | NP_872329.1 | ||
| XM_011533402.2 | XP_011531704.1 | |||
| XM_017005760.1 | XP_016861249.1 | |||
| BC052644.1 | AAH52644.1 | |||
| BX648671.1 | ||||
| CR749370.1 | CAH18223.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893678 | Chr.3:28219843 - 28293336 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map